Gene Transcripts
- Gene Symbol
- ASB18
- Entrez Gene ID
- 401036
- Gene Name
- ankyrin repeat and SOCS box containing 18
- Gene Aliases
- ASB-18
- Chromosome Location
- Chr. 2: 236194872 - 236264345 on Build GRCh38
- UniGene ID
- Hs.639909
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK123854.1 |
- |
1 |
160 |
|
View Full Product Details