Gene Transcripts
- Gene Symbol
- SLC20A2
- Entrez Gene ID
- 6575
- Gene Name
- solute carrier family 20 member 2
- Gene Aliases
- GLVR-2, GLVR2, IBGC1, IBGC3, MLVAR, PIT-2, PIT2, RAM1, Ram-1
- Chromosome Location
- Chr. 8: 42416462 - 42542213 on Build GRCh38
- UniGene ID
- Hs.653173
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001257180.1 |
NP_001244109.1
|
7, 8 |
1636 |
|
NM_001257181.1 |
NP_001244110.1
|
7, 8 |
1267 |
|
NM_006749.4 |
NP_006740.1
|
7, 8 |
1599 |
|
XM_005273613.3 |
XP_005273670.1
|
7, 8 |
1374 |
|
XM_005273615.3 |
XP_005273672.1
|
7, 8 |
1374 |
|
XM_006716390.3 |
XP_006716453.1
|
6, 7 |
1233 |
|
XM_006716391.3 |
XP_006716454.1
|
8, 9 |
4854 |
|
XM_017013748.1 |
XP_016869237.1
|
7, 8 |
1490 |
|
XM_017013749.1 |
XP_016869238.1
|
6, 7 |
4578 |
|
XM_017013750.1 |
XP_016869239.1
|
8, 9 |
1509 |
|
XM_017013751.1 |
XP_016869240.1
|
8, 9 |
1383 |
|
XM_017013752.1 |
XP_016869241.1
|
8, 9 |
1625 |
|
GenBank mRNA |
AK291202.1 |
- |
7, 8 |
1636 |
|
AK295952.1 |
- |
3, 4 |
477 |
|
AK314774.1 |
- |
7, 8 |
1268 |
|
BC028600.2 |
- |
7, 8 |
1311 |
4820601 |
L20852.1 |
- |
6, 7 |
1185 |
|
View Full Product Details