Gene Transcripts

Gene Symbol
ARRDC5
Entrez Gene ID
645432
Gene Name
arrestin domain containing 5
Gene Aliases
Chromosome Location
Chr. 19: 4867614 - 4915469 on Build GRCh38
UniGene ID
Hs.574574
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080523.1 NP_001073992.1 3 661
XM_005259616.2 XP_005259673.1 3 395
XM_011528181.2 XP_011526483.1 3 440

View Full Product Details