Gene Transcripts
- Gene Symbol
- BOLA2
- Entrez Gene ID
- 552900
- Gene Name
- bolA family member 2
- Gene Aliases
- BOLA2A, BOLA2B, My016
- Chromosome Location
- Chr. 16: 29453588 - 29454964 on Build GRCh38
- UniGene ID
- Hs.444600
- Species
- Homo sapiens
- Gene Symbol
- BOLA2-SMG1P6
- Entrez Gene ID
- 107282092
- Gene Name
- BOLA2-SMG1P6 readthrough
- Gene Aliases
- Chromosome Location
- Chr. 16: 29442905 - 29454964 on Build GRCh38
- UniGene ID
- Hs.444600
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK027255.1 |
- |
2 |
590 |
|
- Gene Symbol
- BOLA2B
- Entrez Gene ID
- 654483
- Gene Name
- bolA family member 2B
- Gene Aliases
- BOLA2
- Chromosome Location
- Chr. 16: 30192930 - 30194306 on Build GRCh38
- UniGene ID
- Hs.444600
- Species
- Homo sapiens
View Full Product Details