Gene Transcripts

Gene Symbol
Cxcr3
Entrez Gene ID
12766
Gene Name
chemokine (C-X-C motif) receptor 3
Gene Aliases
Cd183, Cmkar3
Chromosome Location
Chr. X: 101731536 - 101734147 on Build GRCm38
UniGene ID
Mm.12876
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AB003174.1 - 2 229
AF045146.1 - 1 140
BC096626.1 - 2 202 30749408

View Full Product Details