Gene Transcripts

Gene Symbol
Akr1b8
Entrez Gene ID
14187
Gene Name
aldo-keto reductase family 1, member B8
Gene Aliases
FR-1, Fgfrp, Fgrp
Chromosome Location
Chr. 6: 34354164 - 34368454 on Build GRCm38
UniGene ID
Mm.5378
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008012.1 NP_032038.1 4 468
GenBank mRNA AK207771.1 - 1 37
BC005789.1 - 4 457 3603424
U04204.1 - 4 468

View Full Product Details