Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001163282.2 | NP_001156754.1 | 8 | 2541 | |
NM_008107.4 | NP_032133.2 | 2 | 1219 | ||
GenBank mRNA | AK162823.1 | - | 8 | 2524 | |
BC079555.1 | - | 3 | 1470 | 30359238 | |
BC094351.1 | - | 8 | 2493 | ||
M57639.1 | - | 2 | 1219 | ||
M62301.1 | - | 8 | 2529 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_138647.3 | NP_619588.1 | 8 | 2541 | |
GenBank mRNA | AK053885.1 | - | 8 | 2546 |