Gene Transcripts

Gene Symbol
Hmgb3
Entrez Gene ID
15354
Gene Name
high mobility group box 3
Gene Aliases
Hmg2a, Hmg4
Chromosome Location
Chr. X: 71555917 - 71560673 on Build GRCm38
UniGene ID
Mm.336087
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001293623.1 NP_001280552.1 3 528
NM_001293624.1 NP_001280553.1 3 299
NM_001293625.1 NP_001280554.1 3 295
NM_008253.4 NP_032279.1 3 481
GenBank mRNA AF022465.1 - 2 229
AK032163.1 - 3 405
AK141399.1 - 3 298
AK146795.1 - 3 298
AK159119.1 - 3 272
AK159322.1 - 3 272
BC011276.1 - 3 262 3980194
BC083352.1 - 2 213 30540227
BC158037.1 - 3 233 9007340
CX240626.1 - 3 264

View Full Product Details