Gene Transcripts

Gene Symbol
Hmgb3
Entrez Gene ID
15354
Gene Name
high mobility group box 3
Gene Aliases
Hmg2a, Hmg4
Chromosome Location
Chr. X: 71555917 - 71560673 on Build GRCm38
UniGene ID
Mm.336087
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001293623.1 NP_001280552.1 2 402
NM_001293624.1 NP_001280553.1 2 173
NM_001293625.1 NP_001280554.1 2 169
NM_008253.4 NP_032279.1 2 355
GenBank mRNA AF022465.1 - 1 103
AI385779.1 - 2 355
AK032163.1 - 2 279
AK141399.1 - 2 172
AK146795.1 - 2 172
AK159119.1 - 2 146
AK159322.1 - 2 146
BC011276.1 - 2 136 3980194
BC083352.1 - 1 87 30540227
BC158037.1 - 2 107 9007340
CX240626.1 - 2 138

View Full Product Details