Gene Transcripts
- Gene Symbol
- Mtf2
- Entrez Gene ID
- 17765
- Gene Name
- metal response element binding transcription factor 2
- Gene Aliases
- 9230112N11Rik, AA537621, C76717, M96, Pcl2
- Chromosome Location
- Chr. 5: 108065669 - 108109219 on Build GRCm38
- UniGene ID
- Mm.257149
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001253877.1 |
NP_001240806.1
|
9, 10 |
1444 |
|
NM_001253878.1 |
NP_001240807.1
|
9, 10 |
1163 |
|
NM_001253879.1 |
NP_001240808.1
|
8, 9 |
1086 |
|
NM_001253880.1 |
NP_001240809.1
|
6, 7 |
801 |
|
NM_013827.3 |
NP_038855.2
|
8, 9 |
1082 |
|
GenBank mRNA |
AB221622.1 |
- |
5, 6 |
489 |
|
AK030752.1 |
- |
8, 9 |
1032 |
|
AK031409.1 |
- |
10, 11 |
1151 |
|
AK032798.1 |
- |
9, 10 |
1405 |
|
AK032819.1 |
- |
9, 10 |
1406 |
|
AK137539.1 |
- |
10, 11 |
1391 |
|
AK139685.1 |
- |
9, 10 |
1164 |
|
BC024889.1 |
- |
8, 9 |
1054 |
|
BC029076.1 |
- |
8, 9 |
993 |
|
BC050826.1 |
- |
8, 9 |
1036 |
|
BC057163.1 |
- |
8, 9 |
879 |
|
BC071223.1 |
- |
6, 7 |
554 |
|
BC092237.1 |
- |
8, 9 |
1014 |
30637248 |
BC100340.1 |
- |
6, 7 |
732 |
30715034 |
S78454.1 |
- |
5, 6 |
489 |
|
View Full Product Details