Gene Transcripts
- Gene Symbol
- Myl6
- Entrez Gene ID
- 17904
- Gene Name
- myosin, light polypeptide 6, alkali, smooth muscle and non-muscle
- Gene Aliases
- ESMLC, LC17, LC17-GI, LC17-NM, LC17A, LC17B, MLC-3, MLC1SM, MLC3SM, MLC3nm, Myln
- Chromosome Location
- Chr. 10: 128490861 - 128493896 on Build GRCm38
- UniGene ID
- Mm.337074
- Species
- Mus musculus
- Gene Symbol
- Gm5526
- Entrez Gene ID
- 433297
- Gene Name
- myosin, light polypeptide 6, alkali, smooth muscle and non-muscle pseudogene
- Gene Aliases
- EG433297
- Chromosome Location
- Chr. 1: 45857188 - 45857741 on Build GRCm38
- UniGene ID
- Mm.491228
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AA261554.1 |
- |
1 |
148 |
|
View Full Product Details