Gene Transcripts
- Gene Symbol
- Myo5a
- Entrez Gene ID
- 17918
- Gene Name
- myosin VA
- Gene Aliases
- 9630007J19Rik, AI413174, AI661011, Dbv, MVa, Myo5, MyoVA, Sev-1, d, d-120J, flail, flr
- Chromosome Location
- Chr. 9: 75069298 - 75223688 on Build GRCm38
- UniGene ID
- Mm.3645
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_010864.2 |
NP_034994.2
|
32 |
4629 |
|
GenBank mRNA |
X57377.1 |
- |
32 |
4242 |
|
View Full Product Details