Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_010877.4 | NP_035007.1 | 2 | 382 | |
GenBank mRNA | AB002664.1 | - | 2 | 336 | |
AK036379.1 | - | 2 | 383 | ||
AK137152.1 | - | 2 | 437 | ||
AK153447.1 | - | 2 | 344 | ||
AK155324.1 | - | 2 | 437 | ||
AK170839.1 | - | 2 | 407 | ||
AK170998.1 | - | 2 | 410 | ||
AK171059.1 | - | 2 | 408 | ||
AK171265.1 | - | 2 | 410 | ||
BC003730.1 | - | 2 | 330 | 3592460 | |
BC076609.1 | - | 3 | 1336 | ||
BC094056.1 | - | 2 | 428 | ||
DQ449939.1 | - | 2 | 337 |