Gene Transcripts

Gene Symbol
Mapk11
Entrez Gene ID
19094
Gene Name
mitogen-activated protein kinase 11
Gene Aliases
P38b, Prkm11, Sapk2, Sapk2b, p38-2, p38beta, p38beta2
Chromosome Location
Chr. 15: 89142481 - 89149623 on Build GRCm38
UniGene ID
Mm.91969
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011161.5 NP_035291.4 11 958
GenBank mRNA AF135185.1 - 11 938
AK144846.1 - 11 931
AK161190.1 - 9 1133
AK161562.1 - 11 910
BC057211.1 - 11 894
BC064737.1 - 11 909 30112690
BC092526.1 - 11 958 6593321

View Full Product Details