Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001077363.2 | NP_001070831.1 | 4 | 611 | |
NM_001283013.1 | NP_001269942.1 | 4 | 491 | ||
NM_008956.3 | NP_032982.2 | 4 | 611 | ||
GenBank mRNA | AB074764.1 | - | 3 | 329 | |
AK033319.1 | - | 4 | 379 | ||
AK036745.1 | - | 4 | 256 | ||
AK037607.1 | - | 4 | 411 | ||
AK041143.1 | - | 4 | 379 | ||
AK043824.1 | - | 4 | 404 | ||
AK044956.1 | - | 4 | 407 | ||
AK050498.1 | - | 4 | 376 | ||
AK051669.1 | - | 4 | 379 | ||
AK053926.1 | - | 4 | 376 | ||
AK082600.1 | - | 4 | 415 | ||
AK088126.1 | - | 4 | 379 | ||
AK088483.1 | - | 4 | 376 | ||
AK149941.1 | - | 4 | 376 | ||
AK150840.1 | - | 4 | 415 | ||
AK151292.1 | - | 4 | 439 | ||
AK163274.1 | - | 4 | 436 | ||
AK168359.1 | - | 4 | 376 | ||
AK168392.1 | - | 4 | 412 | ||
AK172712.1 | - | 4 | 411 | ||
AK202631.1 | - | 2 | 278 | ||
AK205920.1 | - | 2 | 266 | ||
AK207638.1 | - | 2 | 271 | ||
AK214271.1 | - | 2 | 265 | ||
AK216603.1 | - | 2 | 265 | ||
AK219933.1 | - | 2 | 265 | ||
AK220042.1 | - | 1 | 218 | ||
BC007472.1 | - | 4 | 428 | 3601527 | |
BC028848.1 | - | 4 | 378 | 4022742 | |
BC066210.1 | - | 4 | 388 | 30010932 | |
BC086489.1 | - | 4 | 363 | 30439895 | |
X52101.1 | - | 4 | 363 |