Gene Transcripts

Gene Symbol
Stag2
Entrez Gene ID
20843
Gene Name
stromal antigen 2
Gene Aliases
9230105L23Rik, B230112I07Rik, SA-2, SAP2
Chromosome Location
Chr. X: 42149322 - 42277186 on Build GRCm38
UniGene ID
Mm.290422
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001077712.2 NP_001071180.1 7 925
NM_001290713.1 NP_001277642.1 6 651
NM_021465.4 NP_067440.3 7 785
GenBank mRNA AJ002636.1 - 6 627
AK146718.1 - 7 785
AK168899.1 - 7 785
BC066041.1 - 7 666
CO434048.1 - 5 472

View Full Product Details