Gene Transcripts

Gene Symbol
Avpr1b
Entrez Gene ID
26361
Gene Name
arginine vasopressin receptor 1B
Gene Aliases
AVPR3, V3/V1b, VIBR, VPR3
Chromosome Location
Chr. 1: 131599114 - 131612000 on Build GRCm38
UniGene ID
Mm.89986
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011924.2 NP_036054.1 2 1794
GenBank mRNA AK033416.1 - 2 828
AK050523.1 - 2 876

View Full Product Details