Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_013867.2 | NP_038895.1 | 3 | 742 | |
GenBank mRNA | AF179566.1 | - | 3 | 690 | |
AK143894.1 | - | 3 | 530 | ||
AK158241.1 | - | 3 | 744 | ||
AK167578.1 | - | 3 | 561 | ||
BC023930.1 | - | 3 | 731 | 5324365 |