Gene Transcripts
- Gene Symbol
- Vps37a
- Entrez Gene ID
- 52348
- Gene Name
- vacuolar protein sorting 37A (yeast)
- Gene Aliases
- 2210018P21Rik, 4930592A21Rik, AW261445, D8Ertd531e
- Chromosome Location
- Chr. 8: 40511783 - 40551134 on Build GRCm38
- UniGene ID
- Mm.489696
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_033560.3 |
NP_291038.2
|
5, 6 |
962 |
|
GenBank mRNA |
AK016385.1 |
- |
5, 6 |
945 |
|
AK048942.1 |
- |
5, 6 |
942 |
|
AK054055.1 |
- |
5, 6 |
944 |
|
AK137479.1 |
- |
5, 6 |
947 |
|
AK157507.1 |
- |
5, 6 |
929 |
|
AK170280.1 |
- |
5, 6 |
931 |
|
AK170943.1 |
- |
5, 6 |
920 |
|
AK170961.1 |
- |
5, 6 |
951 |
|
AK171055.1 |
- |
5, 6 |
951 |
|
AK171221.1 |
- |
5, 6 |
963 |
|
BC039290.1 |
- |
5, 6 |
918 |
5066508 |
BC110396.1 |
- |
5, 6 |
877 |
5253683 |
View Full Product Details