Gene Transcripts

Gene Symbol
Hist1h1b
Entrez Gene ID
56702
Gene Name
histone cluster 1, H1b
Gene Aliases
H1.5, H1B, H1f5, H1s-3
Chromosome Location
Chr. 13: 21779832 - 21780625 on Build GRCm38
UniGene ID
Mm.221314
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020034.2 NP_064418.1 1 383
GenBank mRNA AK020117.1 - 1 384
BC114579.1 - 1 311

View Full Product Details