Gene Transcripts

Gene Symbol
Aig1
Entrez Gene ID
66253
Gene Name
androgen-induced 1
Gene Aliases
1500031O19Rik, AV064870, AW413422, CGI-103
Chromosome Location
Chr. 10: 13647054 - 13869036 on Build GRCm38
UniGene ID
Mm.45755
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025446.3 NP_079722.1 3 401
GenBank mRNA AK005332.1 - 3 429
AK008210.1 - 3 404
AK147713.1 - 4 601
AK162306.1 - 3 420
BC035305.1 - 3 551 3980923
EF410113.1 - 1 286

View Full Product Details