Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001162939.1 | NP_001156411.1 | 3 | 728 | |
NM_026531.4 | NP_080807.3 | 3 | 842 | ||
GenBank mRNA | AK012555.1 | - | 3 | 827 | |
AK017644.1 | - | 2 | 187 | ||
AK031972.1 | - | 3 | 843 | ||
AK076421.1 | - | 198 | |||
AK089090.1 | - | 3 | 664 | ||
AK090056.1 | - | 3 | 711 | ||
AK165708.1 | - | 3 | 722 | ||
AK166806.1 | - | 3 | 705 | ||
AK167042.1 | - | 3 | 705 | ||
AK167658.1 | - | 3 | 801 | ||
BC022614.1 | - | 3 | 922 | 4167800 | |
CX219175.1 | - | 2 | 314 |