Gene Transcripts

Gene Symbol
Trnp1
Entrez Gene ID
69539
Gene Name
TMF1-regulated nuclear protein 1
Gene Aliases
2300002D11Rik, Trnp
Chromosome Location
Chr. 4: 133491097 - 133499137 on Build GRCm38
UniGene ID
Mm.151594
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081156.2 NP_001074625.1 2 1230
GenBank mRNA BC049168.1 - 2 1186

View Full Product Details