Gene Transcripts

Gene Symbol
Lrr1
Entrez Gene ID
69706
Gene Name
leucine rich repeat protein 1
Gene Aliases
2410005L11Rik, AU021780, LRR-1, Ppil5
Chromosome Location
Chr. 12: 69168814 - 69179010 on Build GRCm38
UniGene ID
Mm.28847
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081406.1 NP_001074875.1 4 1457
GenBank mRNA AK010419.1 - 3 728
AK144741.1 - 4 1458
BC022648.1 - 3 689
BC075711.1 - 4 1438

View Full Product Details