Gene Transcripts

Gene Symbol
Nup43
Entrez Gene ID
69912
Gene Name
nucleoporin 43
Gene Aliases
2610016K01Rik, 2610529I12Rik, AA409950, p42
Chromosome Location
Chr. 10: 7667504 - 7678886 on Build GRCm38
UniGene ID
Mm.299887
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145706.2 NP_663752.2 8 1082
GenBank mRNA AK011422.1 - 8 1046
AK012191.1 - 8 1076
AK090019.1 - 8 1045
AK146262.1 - 8 1084
BC011060.1 - 8 1070 3156436
BC052530.1 - 8 1010
DV046453.1 - 555

View Full Product Details