Gene Transcripts

Gene Symbol
Armc3
Entrez Gene ID
70882
Gene Name
armadillo repeat containing 3
Gene Aliases
4921513G22Rik
Chromosome Location
Chr. 2: 19198865 - 19310243 on Build GRCm38
UniGene ID
Mm.276696
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081083.2 NP_001074552.1 20 2777
NM_001271563.1 NP_001258492.1 19 2741
NM_001271564.1 NP_001258493.1 19 2729
NM_001271565.1 NP_001258494.1 18 2578
GenBank mRNA AK014880.1 - 20 2593
BC145004.1 - 18 2386
BC145005.1 - 19 2549 9053553
BC145013.1 - 19 2537 9053561
BC150688.1 - 19 2549 9087599

View Full Product Details