Gene Transcripts
- Gene Symbol
- Ccdc57
- Entrez Gene ID
- 71276
- Gene Name
- coiled-coil domain containing 57
- Gene Aliases
- 4933434G05Rik
- Chromosome Location
- Chr. 11: 120826529 - 120932902 on Build GRCm38
- UniGene ID
- Mm.275751
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_027745.1 |
NP_082021.1
|
15 |
2441 |
|
GenBank mRNA |
BC056480.1 |
- |
15 |
2441 |
|
View Full Product Details