Gene Transcripts

Gene Symbol
ACAT2
Entrez Gene ID
39
Gene Name
acetyl-CoA acetyltransferase 2
Gene Aliases
Chromosome Location
Chr. 6: 159761957 - 159779112 on Build GRCh38
UniGene ID
Hs.571037
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001303253.1 NP_001290182.1 7 1080
NM_005891.2 NP_005882.2 7 965
GenBank mRNA AB208993.1 - 7 855
AF356877.1 - 7 888
AK055001.1 - 2 1064
AK225089.1 - 7 866
AK225244.1 - 7 878
AK294273.1 - 7 1080
BC000408.2 - 7 845 2823036
S70154.1 - 7 871

View Full Product Details