Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_028238.7 | NP_082514.4 | 1, 2 | 329 | |
GenBank mRNA | AB232634.1 | - | 1, 2 | 200 | |
AK009296.1 | - | 1, 2 | 290 | ||
AK044783.1 | - | 1, 2 | 292 | ||
AK054253.1 | - | 1, 2 | 330 | ||
AK165682.1 | - | 1, 2 | 332 | ||
AK198599.1 | - | 1, 2 | 87 | ||
AY062237.1 | - | 1, 2 | 289 | ||
BC089578.1 | - | 1, 2 | 289 | 6762755 |