Gene Transcripts

Gene Symbol
MYL6
Entrez Gene ID
4637
Gene Name
myosin light chain 6
Gene Aliases
ESMLC, LC17, LC17-GI, LC17-NM, LC17A, LC17B, MLC-3, MLC1SM, MLC3NM, MLC3SM
Chromosome Location
Chr. 12: 56158261 - 56161582 on Build GRCh38
UniGene ID
Hs.632717
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021019.4 NP_066299.2 4 423
NM_079423.3 NP_524147.2 4 423
GenBank mRNA AK301198.1 - 3 601
BC006781.2 - 2 253
BC017455.1 - 4 310 3538373
BC071661.1 - 4 306 5561575
BC093066.1 - 4 319 30722094
BG283793.1 - 4 422
CN401857.1 - 4 307
CR456839.1 - 2 177
CR457014.1 - 3 282
M22918.1 - 4 313
M22920.1 - 3 291
M31212.1 - 4 322
U02629.1 - 4 320

View Full Product Details