Gene Transcripts

Gene Symbol
Lrrc15
Entrez Gene ID
74488
Gene Name
leucine rich repeat containing 15
Gene Aliases
5430427N11Rik, LIB
Chromosome Location
Chr. 16: 30269302 - 30283254 on Build GRCm38
UniGene ID
Mm.290677
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028973.2 NP_083249.1 2 435
GenBank mRNA AK017350.1 - 2 435
BC050245.1 - 2 406 6312567

View Full Product Details