Gene Transcripts

Gene Symbol
Noc4l
Entrez Gene ID
100608
Gene Name
nucleolar complex associated 4 homolog (S. cerevisiae)
Gene Aliases
AI326906
Chromosome Location
Chr. 5: 110648419 - 110653382 on Build GRCm38
UniGene ID
Mm.29960
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153570.2 NP_705798.2 11 1061
GenBank mRNA AK052601.1 - 11 1061
AK088355.1 - 11 1059
AK090024.1 - 11 1073
AK172310.1 - 11 1040
BC024616.1 - 11 1051 4218310

View Full Product Details