Gene Transcripts

Gene Symbol
Nxph4
Entrez Gene ID
104080
Gene Name
neurexophilin 4
Gene Aliases
1110036M10Rik, AI851051
Chromosome Location
Chr. 10: 127525473 - 127534559 on Build GRCm38
UniGene ID
Mm.437200
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183297.2 NP_899120.2 2 512
GenBank mRNA AK004130.1 - 2 513

View Full Product Details