Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_013464.4 | NP_038492.1 | 3 | 665 | |
GenBank mRNA | AF405560.1 | - | 3 | 299 | |
AF405561.1 | - | 3 | 299 | ||
AF405562.1 | - | 3 | 299 | ||
AF405563.1 | - | 3 | 299 | ||
AF405565.1 | - | 3 | 299 | ||
AF405566.1 | - | 3 | 299 | ||
AF405567.1 | - | 3 | 299 | ||
AF405568.1 | - | 3 | 299 | ||
AF405570.1 | - | 3 | 299 | ||
AK153678.1 | - | 3 | 666 | ||
AK155380.1 | - | 3 | 717 | ||
AY662080.1 | - | 3 | 299 | ||
AY662082.1 | - | 3 | 299 | ||
AY662083.1 | - | 3 | 299 | ||
AY662084.1 | - | 3 | 299 | ||
D38416.1 | - | 3 | 314 |