Gene Transcripts

Gene Symbol
Ift80
Entrez Gene ID
68259
Gene Name
intraflagellar transport 80
Gene Aliases
4921524P20Rik, Wdr56, mKIAA1374
Chromosome Location
Chr. 3: 68892499 - 69004653 on Build GRCm38
UniGene ID
Mm.389451
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026641.2 NP_080917.1 4 526
GenBank mRNA AK042231.1 - 4 451
AK129339.1 - 4 454
AK147305.1 - 3 4053
BC034101.1 - 4 421 4501451

View Full Product Details