Gene Transcripts
- Gene Symbol
- MMP20
- Entrez Gene ID
- 9313
- Gene Name
- matrix metallopeptidase 20
- Gene Aliases
- AI2A2, MMP-20
- Chromosome Location
- Chr. 11: 102576835 - 102625332 on Build GRCh38
- UniGene ID
- Hs.591946
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_004771.3 |
NP_004762.2
|
3 |
508 |
|
GenBank mRNA |
Y12779.1 |
- |
3 |
508 |
|
View Full Product Details