Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001253866.1 | NP_001240795.1 | 10 | 1232 | |
NM_014263.3 | NP_055078.1 | 11 | 1331 | ||
NM_139312.2 | NP_647473.1 | 12 | 1502 | ||
XM_011519300.2 | XP_011517602.1 | 11 | 1785 | ||
GenBank mRNA | AF070656.1 | - | 7 | 646 | |
AF151782.1 | - | 12 | 1346 | ||
AJ132637.1 | - | 11 | 1304 | ||
AK001259.1 | - | 2 | 93 | ||
AK291292.1 | - | 11 | 1260 | ||
AK297973.1 | - | 10 | 1196 | ||
AK309708.1 | - | 7 | 777 | ||
AK314372.1 | - | 11 | 1259 | ||
AY358484.1 | - | 11 | 1257 | ||
BC007795.2 | - | 12 | 1438 | 4110395 | |
BC023507.2 | - | 11 | 1273 | 3637576 | |
BC024032.2 | - | 11 | 1278 | 2961446 |