Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001100409.2 | NP_001093879.1 | 6 | 1530 | |
NM_001304792.1 | NP_001291721.1 | 6 | 1530 | ||
NM_015571.3 | NP_056386.2 | 7 | 1551 | ||
XM_006715457.3 | XP_006715520.1 | 8 | 1305 | ||
XM_011535746.2 | XP_011534048.1 | 6 | 1103 | ||
XM_011535747.2 | XP_011534049.1 | 6 | 1454 | ||
XM_017010719.1 | XP_016866208.1 | 7 | 1193 | ||
XM_017010720.1 | XP_016866209.1 | 7 | 509 | ||
XM_017010721.1 | XP_016866210.1 | 7 | 517 | ||
XM_017010722.1 | XP_016866211.1 | 8 | 1324 | ||
XM_017010723.1 | XP_016866212.1 | 8 | 1301 | ||
XM_017010724.1 | XP_016866213.1 | 8 | 1676 | ||
XM_017010725.1 | XP_016866214.1 | 5 | 1436 | ||
GenBank mRNA | AB018340.2 | - | 7 | 579 | |
AF196304.1 | - | 7 | 535 | ||
AF306508.1 | - | 7 | 550 | ||
AF307849.1 | - | 7 | 610 | ||
AK001406.1 | - | 4 | 205 | ||
AK096455.1 | - | 6 | 1112 | ||
AK126921.1 | - | 6 | 1530 | ||
BC028583.1 | - | 6 | 737 | 4838569 |