Gene Transcripts

Gene Symbol
HOXC10
Entrez Gene ID
3226
Gene Name
homeobox C10
Gene Aliases
HOX3I
Chromosome Location
Chr. 12: 53985162 - 53990279 on Build GRCh38
UniGene ID
Hs.44276
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017409.3 NP_059105.2 1 427
GenBank mRNA AF255675.1 - 1 329
AK313759.1 - 1 443
AL558161.3 - 1 427
BC001293.1 - 1 400 3458115
BT009921.1 - 1 329

View Full Product Details