Gene Transcripts
- Gene Symbol
- SLC29A3
- Entrez Gene ID
- 55315
- Gene Name
- solute carrier family 29 member 3
- Gene Aliases
- ENT3, HCLAP, HJCD, PHID
- Chromosome Location
- Chr. 10: 71319253 - 71363390 on Build GRCh38
- UniGene ID
- Hs.438419
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_018344.5 |
NP_060814.4
|
5, 6 |
838 |
|
NR_033413.1 |
|
4, 5 |
755 |
|
NR_033414.1 |
|
3, 4 |
528 |
|
XM_006717910.3 |
XP_006717973.1
|
5, 6 |
1230 |
|
XM_017016377.1 |
XP_016871866.1
|
4, 5 |
1052 |
|
XM_017016378.1 |
XP_016871867.1
|
3, 4 |
828 |
|
GenBank mRNA |
AF326987.1 |
- |
4, 5 |
786 |
|
AK002022.1 |
- |
4, 5 |
589 |
|
AK304503.1 |
- |
4, 5 |
725 |
|
AK314497.1 |
- |
5, 6 |
806 |
|
AK316152.1 |
- |
4, 5 |
740 |
|
AX775915.1 |
- |
5, 6 |
838 |
|
AX775917.1 |
- |
5, 6 |
838 |
|
AX775919.1 |
- |
5, 6 |
838 |
|
AX775921.1 |
- |
5, 6 |
838 |
|
AY288928.1 |
- |
5, 6 |
798 |
|
AY358686.1 |
- |
5, 6 |
816 |
|
BC000223.1 |
- |
2, 3 |
213 |
|
BC041575.1 |
- |
4, 5 |
726 |
|
BC120996.1 |
- |
5, 6 |
816 |
40116513 |
BC120997.1 |
- |
5, 6 |
816 |
40116514 |
View Full Product Details