Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000277.1 | NP_000268.1 | 12 | 1767 | |
GenBank mRNA | AK313383.1 | - | 13 | 1392 | |
BC026251.1 | - | 12 | 1448 | 4767736 | |
K03020.1 | - | 12 | 1517 | ||
L47726.1 | - | 12 | 1767 | ||
U49897.1 | - | 12 | 1767 |