Gene Transcripts

Gene Symbol
CAPN15
Entrez Gene ID
6650
Gene Name
calpain 15
Gene Aliases
SOLH
Chromosome Location
Chr. 16: 527795 - 554636 on Build GRCh38
UniGene ID
Hs.632219
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005632.2 NP_005623.1 14 4616
XM_011522620.2 XP_011520922.1 15 7939
XM_011522621.2 XP_011520923.1 15 8867
XM_011522622.1 XP_011520924.1 13 7571
XM_011522623.2 XP_011520925.1 16 5642
XM_011522624.2 XP_011520926.1 15 4980
XM_011522625.2 XP_011520927.1 14 4866
XM_011522626.2 XP_011520928.1 15 8407
XM_011522627.2 XP_011520929.1 15 8408
XM_011522628.2 XP_011520930.1 15 5500
XM_011522629.2 XP_011520931.1 15 5363
XM_011522630.2 XP_011520932.1 14 4701
XM_011522631.2 XP_011520933.1 13 4587
XM_011522632.2 XP_011520934.1 13 5018
XM_017023596.1 XP_016879085.1 16 5779
GenBank mRNA BC001869.1 - 2 1342
BC021854.1 - 10 2683
BC040169.1 - 1 812
BC112918.1 - 6 2841
U69557.1 - 2 1302

View Full Product Details