Gene Transcripts

Gene Symbol
MARCKS
Entrez Gene ID
4082
Gene Name
myristoylated alanine rich protein kinase C substrate
Gene Aliases
80K-L, MACS, PKCSL, PRKCSL
Chromosome Location
Chr. 6: 113857335 - 113863475 on Build GRCh38
UniGene ID
Hs.519909
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002356.6 NP_002347.5 2 1208
GenBank mRNA BC089040.1 - 2 1055
D10522.1 - 2 1166
M68956.1 - 2 1105

View Full Product Details