Gene Transcripts

Gene Symbol
MMP21
Entrez Gene ID
118856
Gene Name
matrix metallopeptidase 21
Gene Aliases
HTX7, MMP-21
Chromosome Location
Chr. 10: 125766453 - 125775821 on Build GRCh38
UniGene ID
Hs.314141
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_147191.1 NP_671724.1 6 1379
GenBank mRNA AF331526.1 - 6 1379
AF520613.1 - 6 1379

View Full Product Details