Gene Transcripts

Gene Symbol
USF1
Entrez Gene ID
7391
Gene Name
upstream transcription factor 1
Gene Aliases
FCHL, FCHL1, HYPLIP1, MLTF, MLTFI, UEF, bHLHb11
Chromosome Location
Chr. 1: 161039251 - 161045979 on Build GRCh38
UniGene ID
Hs.414880
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001276373.1 NP_001263302.1 6 460
NM_007122.4 NP_009053.1 6 556
NM_207005.2 NP_996888.1 6 525
GenBank mRNA AB098540.1 - 4 318
AK314876.1 - 6 476
AL832119.1 - 6 593
BC035505.2 - 6 504 4559819
KU178379.1 - 4 349
KU178380.1 - 4 318
X55666.1 - 6 469

View Full Product Details