Gene Transcripts

Gene Symbol
IRX4
Entrez Gene ID
50805
Gene Name
iroquois homeobox 4
Gene Aliases
IRXA3
Chromosome Location
Chr. 5: 1877427 - 1887179 on Build GRCh38
UniGene ID
Hs.196927
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278632.1 NP_001265561.1 6 994
NM_001278633.1 NP_001265562.1 7 1072
NM_001278634.1 NP_001265563.1 6 999
NM_001278635.1 NP_001265564.1 7 1077
NM_016358.2 NP_057442.1 5 895
GenBank mRNA AB690778.1 - 6 994
AB690779.1 - 7 1072
AB690780.1 - 6 999
AB690781.1 - 7 1077
AF124733.1 - 5 895
BC110912.1 - 5 783 3450161
BC136505.1 - 5 883 9020494
BC136506.1 - 5 883 9020495

View Full Product Details