Gene Transcripts

Gene Symbol
RING1
Entrez Gene ID
6015
Gene Name
ring finger protein 1
Gene Aliases
RING1A, RNF1
Chromosome Location
Chr. 6: 33208509 - 33212722 on Build GRCh38
UniGene ID
Hs.631989
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002931.3 NP_002922.2 7 1524
GenBank mRNA AK289355.1 - 7 1533
AK301378.1 - 7 1467
BC002922.2 - 7 1519 3954313
BC051866.2 - 7 1517 6142438
Z14000.1 - 5 1318

View Full Product Details