Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001324400.1 | NP_001311329.1 | 5 | 719 | |
NM_022444.3 | NP_071889.2 | 4 | 563 | ||
XM_011516515.2 | XP_011514817.1 | 4 | 567 | ||
XM_011516516.2 | XP_011514818.1 | 4 | 567 | ||
XM_011516517.2 | XP_011514819.1 | 5 | 686 | ||
XM_011516518.2 | XP_011514820.1 | 4 | 567 | ||
XM_011516519.2 | XP_011514821.1 | 4 | 567 | ||
XM_017012554.1 | XP_016868043.1 | 3 | 453 | ||
XM_017012555.1 | XP_016868044.1 | 4 | 567 | ||
GenBank mRNA | AF260824.1 | - | 4 | 563 | |
AK026413.1 | - | 4 | 561 | ||
AK314537.1 | - | 4 | 561 | ||
BC111775.1 | - | 5 | 719 |