Gene Transcripts

Gene Symbol
RAP2A
Entrez Gene ID
5911
Gene Name
RAP2A, member of RAS oncogene family
Gene Aliases
K-REV, KREV, RAP2, RbBP-30
Chromosome Location
Chr. 13: 97434221 - 97467998 on Build GRCh38
UniGene ID
Hs.508480
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021033.6 NP_066361.1 2 944
GenBank mRNA AK311202.1 - 2 453
BC041333.2 - 2 945 5278625
BC070031.1 - 2 702 4815530

View Full Product Details