Gene Transcripts

Gene Symbol
PSMB9
Entrez Gene ID
5698
Gene Name
proteasome subunit beta 9
Gene Aliases
LMP2, PSMB6i, RING12, beta1i
Chromosome Location
Chr. 6: 32854161 - 32859851 on Build GRCh38
UniGene ID
Hs.654585
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002800.4 NP_002791.1 4, 5 459
GenBank mRNA AK303118.1 - 3, 4 330
BC065513.1 - 4, 5 414 5749985
BI832878.1 - 4, 5 441
BX641100.1 - 2, 3 2140
CR541656.1 - 4, 5 390
S75169.1 - 3, 4 369
U01025.1 - 3, 4 369
X62741.1 - 4, 5 405

View Full Product Details